Learn More
Arcticwhite LLC ArctiSeal 96 Well Round, White Silicone, PTFE Coating for Tube Inserts AWLS-T110-2, Pre-Slit, for Manual and Automated Sampling, 5 Sealing Mats per Pack
SDP
Catalog No. NC9243120
Encompass
ArctiSeal 96 Well Round, White Silicone Sealing Mat w/ PTFE Coating for Tube Inserts, Pre-Slit, for Manual and Automated Sampling, 5 Sealing per Pack. This silicone sealing mat seals AWLS-T110-2 96 Well Plate and PCR plates. Supplied by ArcticWhite as 5 sealing mats per pack.
Catalog No. NC9243120 Supplier Arcticwhite LLC Supplier No. AWSM2001RTX
ArctiSeal 96 Well Round, White Silicone Sealing Mat w/ PTFE Coating for Tube Inserts, Pre-Slit, for Manual and Automated Sampling, 5 Sealing per Pack. This silicone sealing mat seals AWLS-T110-2 96 Well Plate and PCR plates. Supplied by ArcticWhite as 5 sealing mats per pack.