Learn More
Enzo Life Sciences Arcyriaflavin A (1mg). CAS: 118458-54-1
SDP
Supplier:  Enzo Life Sciences ALX350375M001
Encompass_Preferred
Potent inhibitor of CDK4 / cyclin D1 (IC50 = 59nM). Inhibits human cytomegalovirus (HCMV) replication in cell culture (IC50 = 200nM). Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8870