Learn More
Medchemexpress LLC ARD-2051 | 2632305-17-8 | 98.6% | 789.32 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY149862
Encompass_Preferred
ARD-2051 is a selective and orally active androgen receptor (AR) proteolysis-targeting chimera degrader. It achieves DC50 values of 0.6 nM for AR protein degradation in both LNCaP and VCaP prostate cancer cell lines, and shows effective anti-tumor activity in a VCaP xenograft mice model. It can be used for the research of prostate cancer.
  • Induces AR protein degradation in LNCaP and VCaP cells with a DC50 of 0.6 nM.
  • Inhibits cell growth in LNCaP and VCaP cells with an IC50 of 12.8 nM and 10.2 nM respectively.
  • Dose-dependently suppresses the expression of AR-regulated gene KLK3.
  • Exhibits effective anti-tumor activity in VCaP xenograft mice model.
  • Achieved tumor growth inhibition (TGI) rates up to 80% at 25 mg/kg in VCaP xenograft mice.
Catalog No. 50-003-61297