Learn More
Medchemexpress LLC Ard-2051 | 2632305-17-8 | 98.6% | 789.32 | C43H45ClN8O5 | 10 MG
SDP
Catalog No. 5000287344
Encompass_Preferred
ARD-2051 is a selective, orally active androgen receptor proteolysis-targeting chimera (PROTAC) degrader developed for preclinical research into AR-dependent cancers. It induces potent AR protein degradation in cell-based assays and has demonstrated favorable pharmacokinetics and oral efficacy in animal models.
  • Potent AR protein degradation (DC50 ~0.6 nM).
  • Orally active with demonstrated in vivo efficacy in preclinical models.
  • Effective in AR-positive prostate cancer cell lines, including LNCaP and VCaP.
  • Characterized pharmacokinetic profile and oral bioavailability in rodent and canine studies.
  • High chemical purity suitable for research applications.
  • Supplied as a solid with defined storage recommendations for stability.
Catalog No. 50-002-87344 Supplier Medchemexpress LLC Supplier No. HY14986210MG
ARD-2051 is a selective, orally active androgen receptor proteolysis-targeting chimera (PROTAC) degrader developed for preclinical research into AR-dependent cancers. It induces potent AR protein degradation in cell-based assays and has demonstrated favorable pharmacokinetics and oral efficacy in animal models.
  • Potent AR protein degradation (DC50 ~0.6 nM).
  • Orally active with demonstrated in vivo efficacy in preclinical models.
  • Effective in AR-positive prostate cancer cell lines, including LNCaP and VCaP.
  • Characterized pharmacokinetic profile and oral bioavailability in rodent and canine studies.
  • High chemical purity suitable for research applications.
  • Supplied as a solid with defined storage recommendations for stability.