Learn More
TARGETMOL CHEMICALS INC ARD-2128 1MG
SDP
Catalog No. 502877195
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ARD-2128 is a highly potent, orally bioavailable PROTAC (proteolysis-targeting chimera) that degrades the androgen receptor (AR), effectively reducing AR protein levels, suppressing AR-regulated gene expression in tumor tissues, and inhibiting tumor growth without evident toxicity. This compound holds promise for prostate cancer research. Purity 99.72%
Catalog No. 50-287-7195 Supplier TARGETMOL CHEMICALS INC Supplier No. T396951MG
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ARD-2128 is a highly potent, orally bioavailable PROTAC (proteolysis-targeting chimera) that degrades the androgen receptor (AR), effectively reducing AR protein levels, suppressing AR-regulated gene expression in tumor tissues, and inhibiting tumor growth without evident toxicity. This compound holds promise for prostate cancer research. Purity 99.72%