Learn More
Medchemexpress LLC HY-132292 5mg , ARD-2128 CAS:2222111-87-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1322925MG
Encompass_Preferred
HY-132292 5mg ARD-2128 CAS: 2222111-87-5 ARD-2128 is a highly potent, orally bioavailable PROTAC androgen receptor (AR) degrader. ARD-2128 effectively reduces AR protein, suppresses AR-regulated genes in tumor tissues, and inhibits growth of tumor without signs of toxicity. ARD-2128 has the potential for the research of the prostate cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0723