Learn More
Medchemexpress LLC Ard-2585 | 2757422-79-8 | 98.19% | 763.28 | 25 MG
SDP
Catalog No. 5000348086
Encompass_Preferred
ARD-2585 is described as an exceptionally potent and orally active PROTAC degrader of the androgen receptor.
  • Achieves DC50 values of 0.04-0.1 nM in the VCaP cell line with AR gene amplification and in the LNCaP cell line carrying an AR mutation.
  • At 100 nM, ARD-2585 reduces AR protein level by 78% at 3 hours and 60% at 6 hours.
Catalog No. 50-003-48086 Supplier Medchemexpress LLC Supplier No. 25MGHY139436
ARD-2585 is described as an exceptionally potent and orally active PROTAC degrader of the androgen receptor.
  • Achieves DC50 values of 0.04-0.1 nM in the VCaP cell line with AR gene amplification and in the LNCaP cell line carrying an AR mutation.
  • At 100 nM, ARD-2585 reduces AR protein level by 78% at 3 hours and 60% at 6 hours.