Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ARD-2585 | 2757422-79-8 | 98.19% | 763.28 | 50 MG
SDP

Catalog No. 5000348872
Encompass_Preferred

ARD-2585 is an exceptionally potent and orally active PROTAC degrader of the androgen receptor, intended for research use only. It achieves DC50 values of 0.04-0.1 nM in the VCaP cell line with AR gene amplification and in the LNCaP cell line carrying an AR mutation. ARD-2585 (100 nM) is effective in reducing the AR protein level by 78% at 3 hours and 60% at 6 hours.

  • Purity: 98.19%
  • Molecular weight: 763.28
  • Formula: C41H43ClN8O5
  • Appearance: Solid
  • Color: Light yellow to green yellow
  • Cellular effect: Achieves DC50 values of 0.04-0.1 nM in VCaP and LNCaP cell lines.

Catalog No. 50-003-48872 Supplier Medchemexpress LLC Supplier No. HY13943650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARD-2585 is an exceptionally potent and orally active PROTAC degrader of the androgen receptor, intended for research use only. It achieves DC50 values of 0.04-0.1 nM in the VCaP cell line with AR gene amplification and in the LNCaP cell line carrying an AR mutation. ARD-2585 (100 nM) is effective in reducing the AR protein level by 78% at 3 hours and 60% at 6 hours.

  • Purity: 98.19%
  • Molecular weight: 763.28
  • Formula: C41H43ClN8O5
  • Appearance: Solid
  • Color: Light yellow to green yellow
  • Cellular effect: Achieves DC50 values of 0.04-0.1 nM in VCaP and LNCaP cell lines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.