Learn More
Medchemexpress LLC ARD-2585 | 2757422-79-8 | 98.2% | 763.28 | 100 MG
SDP
Catalog No. 5000349016
Encompass_Preferred
ARD-2585 is an exceptionally potent and orally active PROTAC degrader of androgen receptor. This product is for research use only and not sold to patients.
  • Exceptionally potent and orally active PROTAC degrader of androgen receptor.
  • Achieves DC50 values of 0.04-0.1 nM in VCaP and LNCaP cell lines.
  • Reduces androgen receptor protein level by 78% at 3 hours and 60% at 6 hours at 100 nM.
  • Available as a solid, appearing light yellow to green yellow.
  • Soluble in DMSO at 85 mg/mL (111.36 mM).
Catalog No. 50-003-49016 Supplier Medchemexpress LLC Supplier No. HY139436100MG
ARD-2585 is an exceptionally potent and orally active PROTAC degrader of androgen receptor. This product is for research use only and not sold to patients.
  • Exceptionally potent and orally active PROTAC degrader of androgen receptor.
  • Achieves DC50 values of 0.04-0.1 nM in VCaP and LNCaP cell lines.
  • Reduces androgen receptor protein level by 78% at 3 hours and 60% at 6 hours at 100 nM.
  • Available as a solid, appearing light yellow to green yellow.
  • Soluble in DMSO at 85 mg/mL (111.36 mM).