Learn More
Medchemexpress LLC Ard-2585 | 2757422-79-8 | 99.2% | C41H43ClN8O5 | 10MG
SDP
Catalog No. 5000261247
Encompass_Preferred
ARD-2585 is a potent, orally active proteolysis-targeting chimera (PROTAC) developed for preclinical research into androgen receptor-driven diseases such as advanced prostate cancer. It reduces androgen receptor protein levels in cellular assays and includes reported solubility and storage data suitable for laboratory handling; for research use only, not for human administration.
  • Potent, orally active degrader with demonstrated cellular efficacy.
  • Mechanism of action: PROTAC-mediated target degradation of the androgen receptor.
  • Characterized physicochemical properties, including molecular formula and molecular weight.
  • High solubility in DMSO and formulatable in aqueous vehicles for in vivo studies.
  • Stable under recommended laboratory storage conditions (sealed, protected from moisture and light).
Catalog No. 50-002-61247 Supplier Medchemexpress LLC Supplier No. HY13943610MG
ARD-2585 is a potent, orally active proteolysis-targeting chimera (PROTAC) developed for preclinical research into androgen receptor-driven diseases such as advanced prostate cancer. It reduces androgen receptor protein levels in cellular assays and includes reported solubility and storage data suitable for laboratory handling; for research use only, not for human administration.
  • Potent, orally active degrader with demonstrated cellular efficacy.
  • Mechanism of action: PROTAC-mediated target degradation of the androgen receptor.
  • Characterized physicochemical properties, including molecular formula and molecular weight.
  • High solubility in DMSO and formulatable in aqueous vehicles for in vivo studies.
  • Stable under recommended laboratory storage conditions (sealed, protected from moisture and light).