Learn More
Medchemexpress LLC HY-139436 5mg , ARD-2585 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1394365MG
Encompass_Preferred
HY-139436 5mg ARD-2585 CAS: ARD-2585 is an exceptionally potent and orally active PROTAC degrader of androgen receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0831