Learn More
Medchemexpress LLC ARD-61 | 2316837-08-6 | 99.6% | 1095.78 | 25 MG
SDP
Catalog No. 5000349033
Encompass_Preferred
ARD-61 is a potent PROTAC androgen receptor (AR) degrader. It effectively degrades AR and progesterone receptors (PR) in AR+ cancer cell lines, induces apoptosis, and inhibits tumor growth in xenograft models. It also functions as a click chemistry reagent with an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Degrades androgen receptor (AR) and progesterone receptors (PR).
  • Induces apoptosis in cancer cells.
  • Inhibits tumor growth in xenograft models.
  • Click chemistry reagent with an alkyne group for CuAAc.
  • Induces G2/M cell cycle arrest.
  • Inhibits Wnt/β-catenin and MYC pathways.
  • Decreases HER2 and HER3 proteins (phosphorylated and un-phosphorylated).
Catalog No. 50-003-49033 Supplier Medchemexpress LLC Supplier No. 25MGHY139659
ARD-61 is a potent PROTAC androgen receptor (AR) degrader. It effectively degrades AR and progesterone receptors (PR) in AR+ cancer cell lines, induces apoptosis, and inhibits tumor growth in xenograft models. It also functions as a click chemistry reagent with an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Degrades androgen receptor (AR) and progesterone receptors (PR).
  • Induces apoptosis in cancer cells.
  • Inhibits tumor growth in xenograft models.
  • Click chemistry reagent with an alkyne group for CuAAc.
  • Induces G2/M cell cycle arrest.
  • Inhibits Wnt/β-catenin and MYC pathways.
  • Decreases HER2 and HER3 proteins (phosphorylated and un-phosphorylated).