Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ard-61 | 2316837-08-6 | 99.6% | 1095.78 | C61H71ClN8O7S | 5 MG
SDP

Catalog No. 5000223146
Encompass_Preferred

ARD-61 is a PROTAC that selectively degrades the androgen receptor (AR) and progesterone receptor (PR), inducing apoptosis in AR-positive cancer cell lines and showing tumor growth inhibition in xenograft models. It includes an alkyne functionality for click chemistry and is provided as a research-grade solid.

  • Selective degradation of androgen and progesterone receptors.
  • PROTAC mechanism engaging VHL E3 ligase to induce target proteolysis.
  • Alkyne functional group compatible with click chemistry (CuAAC).
  • High purity suitable for research (≈99.6%).
  • Soluble in DMSO at 100 mg/mL; formulations available for in vivo dosing.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Catalog No. 50-002-23146 Supplier Medchemexpress LLC Supplier No. HY1396595MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARD-61 is a PROTAC that selectively degrades the androgen receptor (AR) and progesterone receptor (PR), inducing apoptosis in AR-positive cancer cell lines and showing tumor growth inhibition in xenograft models. It includes an alkyne functionality for click chemistry and is provided as a research-grade solid.

  • Selective degradation of androgen and progesterone receptors.
  • PROTAC mechanism engaging VHL E3 ligase to induce target proteolysis.
  • Alkyne functional group compatible with click chemistry (CuAAC).
  • High purity suitable for research (≈99.6%).
  • Soluble in DMSO at 100 mg/mL; formulations available for in vivo dosing.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.