Learn More
Medchemexpress LLC Ard-61 | 2316837-08-6 | 99.6% | C61H71ClN8O7S | 10MG
SDP
Catalog No. 5000260369
Encompass_Preferred
ARD-61 is a PROTAC small molecule that selectively induces proteasomal degradation of the androgen receptor (AR). Supplied for research use only, it is used in cellular and in vivo studies to probe AR biology, apoptosis, and tumor growth inhibition. The supplier provides recommended solubility and formulation protocols for both in vitro and in vivo applications.
  • Highly potent and selective androgen receptor degrader.
  • High purity suitable for pharmacology and cell biology studies.
  • Soluble in DMSO at high concentrations for in vitro assays.
  • Includes recommended in vivo formulation protocols for dosing.
  • Available in multiple package sizes to support different study scales.
Catalog No. 50-002-60369 Supplier Medchemexpress LLC Supplier No. HY13965910MG
ARD-61 is a PROTAC small molecule that selectively induces proteasomal degradation of the androgen receptor (AR). Supplied for research use only, it is used in cellular and in vivo studies to probe AR biology, apoptosis, and tumor growth inhibition. The supplier provides recommended solubility and formulation protocols for both in vitro and in vivo applications.
  • Highly potent and selective androgen receptor degrader.
  • High purity suitable for pharmacology and cell biology studies.
  • Soluble in DMSO at high concentrations for in vitro assays.
  • Includes recommended in vivo formulation protocols for dosing.
  • Available in multiple package sizes to support different study scales.