Learn More
Medchemexpress LLC ARD-69 50mg | 2316837-10-0 | 50 MG
SDP
Catalog No. 5000307766
Encompass_Preferred
ARD-69 is a PROTAC small-molecule degrader that targets the androgen receptor (AR) and is used in preclinical research to investigate AR signaling and castration-resistant prostate cancer. It recruits the VHL E3 ligase to promote ubiquitination and proteasomal degradation of AR, resulting in decreased AR-driven transcription and inhibited proliferation in AR-positive cell lines.
  • Used for preclinical studies of AR degradation and prostate cancer models.
  • Potent AR degrader with low-nanomolar cellular activity.
  • High chemical purity suitable for biological assays.
  • Soluble in DMSO at ≥ 200 mg/mL for in vitro formulation.
  • Stable under recommended storage when protected from light.
Catalog No. 50-003-07766 Supplier Medchemexpress LLC Supplier No. HY11440250MG
Add to Cart
Add to Cart
Request a Quote
ARD-69 is a PROTAC small-molecule degrader that targets the androgen receptor (AR) and is used in preclinical research to investigate AR signaling and castration-resistant prostate cancer. It recruits the VHL E3 ligase to promote ubiquitination and proteasomal degradation of AR, resulting in decreased AR-driven transcription and inhibited proliferation in AR-positive cell lines.
  • Used for preclinical studies of AR degradation and prostate cancer models.
  • Potent AR degrader with low-nanomolar cellular activity.
  • High chemical purity suitable for biological assays.
  • Soluble in DMSO at ≥ 200 mg/mL for in vitro formulation.
  • Stable under recommended storage when protected from light.