Learn More
BPS Bioscience Inc ARE LUCIFERAS RPT LENT 2X500UL
SDP
Catalog No. NC1867100
Encompass
The ARE Luciferase Reporter Lentivirus are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to be transduced into almost all types of mammalian cells, including primary and non-dividing cells. The particles contain a firefly luciferase gene driven by ARE located upstream of the minimal TATA promoter (Figure 1). After transduction, activation of the Nrf2 antioxidant response pathway in the target cells can be monitored by measuring the luciferase activity.Our products are for research use only, not for diagnostic or therapeutic use.BPS BIOSCIENCE INC mfg# 79869
Catalog No. NC1867100 Supplier BPS Bioscience Inc Supplier No. 79869
The ARE Luciferase Reporter Lentivirus are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to be transduced into almost all types of mammalian cells, including primary and non-dividing cells. The particles contain a firefly luciferase gene driven by ARE located upstream of the minimal TATA promoter (Figure 1). After transduction, activation of the Nrf2 antioxidant response pathway in the target cells can be monitored by measuring the luciferase activity.Our products are for research use only, not for diagnostic or therapeutic use.BPS BIOSCIENCE INC mfg# 79869