Learn More
BPS Bioscience Inc ARE Luciferase Reporter HepG2 Cell Line, part number 60513, firefly luciferase, adherent cell line, >1 x 10^6 cells in 1 ml Cell Freezing Medium, validated for Nrf2 pathway response
SDP
Catalog No. NC2964332
Encompass
The BPS Bioscience ARE Luciferase Reporter HepG2 Cell Line (Nrf2 Antioxidant Pathway) is specifically engineered to enable precise monitoring of the Nrf2 antioxidant response pathway. This stable, adherent HepG2 cell line incorporates a firefly luciferase reporter gene under the control of an Antioxidant Response Element (ARE), facilitating the direct correlation between luciferase expression and Nrf2 activation. Its reliable performance has been validated through stimulation with various compounds known to influence the antioxidant pathway.
  • Stable, adherent cell line
  • Features firefly luciferase as the reporter gene
  • Monitors Nrf2 antioxidant response pathway activity
  • Validated with tert-butylhydroquinone, sulforaphane, RTA-408
  • Absence of mycoplasma confirmed
  • Suitable for research applications
  • Stored in liquid nitrogen upon receipt
  • Biosafety Level: BSL-1
Catalog No. NC2964332 Supplier BPS Bioscience Inc Supplier No. 60513
The BPS Bioscience ARE Luciferase Reporter HepG2 Cell Line (Nrf2 Antioxidant Pathway) is specifically engineered to enable precise monitoring of the Nrf2 antioxidant response pathway. This stable, adherent HepG2 cell line incorporates a firefly luciferase reporter gene under the control of an Antioxidant Response Element (ARE), facilitating the direct correlation between luciferase expression and Nrf2 activation. Its reliable performance has been validated through stimulation with various compounds known to influence the antioxidant pathway.
  • Stable, adherent cell line
  • Features firefly luciferase as the reporter gene
  • Monitors Nrf2 antioxidant response pathway activity
  • Validated with tert-butylhydroquinone, sulforaphane, RTA-408
  • Absence of mycoplasma confirmed
  • Suitable for research applications
  • Stored in liquid nitrogen upon receipt
  • Biosafety Level: BSL-1