Learn More
Aviva Systems Biology Corp AREB6 Antibody - N-terminal region (P100657_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100657_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AREB6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AREB6. Alternate names: AREB6, BZP, MGC133261, NIL-2-A, NIL-2A, TCF8, ZEB, ZFHEP, ZFHX1A, FECD6, NIL2A, PPCD3, DELTAEF1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53800