Learn More
Cayman Chemical ArecaIdIn but2ynyl ester 10mg
SDP
Catalog No. 502942211
Encompass_Preferred
A muscarinic acetylcholine receptor agonist; binds to M1, M2, and M3 mChARs (pD2s = 7.93, 8.33, and 7.67, respectively); inhibits calcium channel currents in a whole-cell patch clamp assay using primary rat SPG cells (EC50 = 3.6 µM); inhibits transmural nerve stimulation-induced vasodilation in isolated and washed porcine basilar arterial rings (EC50 = 0.5 µM); reduces MAP and BPM in normotensive rats in a dose-dependent manner
Catalog No. 50-294-2211 Supplier Cayman Chemical Supplier No. 3981610 MG
A muscarinic acetylcholine receptor agonist; binds to M1, M2, and M3 mChARs (pD2s = 7.93, 8.33, and 7.67, respectively); inhibits calcium channel currents in a whole-cell patch clamp assay using primary rat SPG cells (EC50 = 3.6 µM); inhibits transmural nerve stimulation-induced vasodilation in isolated and washed porcine basilar arterial rings (EC50 = 0.5 µM); reduces MAP and BPM in normotensive rats in a dose-dependent manner