Learn More
Medchemexpress LLC Arecaidine but-2-yny 10mg | 119630-77-2 | 10MG
SDP
Catalog No. 5000242168
Encompass_Preferred
Arecaidine but-2-ynyl ester tosylate (ABET) is a selective mAChR M2 agonist that dose-dependently decreases mean arterial pressure and heart rate in rats Arecaidine but-2-ynyl ester tosylate can be used for cardiovascular disease research[1 [2 Arecaidine but-2-ynyl ester (tosylate) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-42168 Supplier Medchemexpress LLC Supplier No. HY10765510MG
Arecaidine but-2-ynyl ester tosylate (ABET) is a selective mAChR M2 agonist that dose-dependently decreases mean arterial pressure and heart rate in rats Arecaidine but-2-ynyl ester tosylate can be used for cardiovascular disease research[1 [2 Arecaidine but-2-ynyl ester (tosylate) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups