Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arecaidine but-2-ynyl ester tosylate | 119630-77-2 | 100.0% | 365.44 g/mol | C18H23NO5S | 5 MG
SDP

Catalog No. 5000240117
Encompass_Preferred

Arecaidine but-2-ynyl ester tosylate is a research-grade muscarinic acetylcholine receptor (mAChR) M2 agonist provided as the tosylate salt. It is used in preclinical pharmacology to probe M2 receptor activity and to study cardiovascular responses such as decreases in mean arterial pressure and heart rate in animal models.

  • High purity of 99.95%.
  • Molecular weight 365.44 g/mol.
  • Molecular formula C18H23NO5S.
  • Supplied as a tosylate salt for enhanced stability.
  • Available in small research quantities suitable for in vitro and in vivo studies.

Catalog No. 50-002-40117 Supplier Medchemexpress LLC Supplier No. HY1076555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arecaidine but-2-ynyl ester tosylate is a research-grade muscarinic acetylcholine receptor (mAChR) M2 agonist provided as the tosylate salt. It is used in preclinical pharmacology to probe M2 receptor activity and to study cardiovascular responses such as decreases in mean arterial pressure and heart rate in animal models.

  • High purity of 99.95%.
  • Molecular weight 365.44 g/mol.
  • Molecular formula C18H23NO5S.
  • Supplied as a tosylate salt for enhanced stability.
  • Available in small research quantities suitable for in vitro and in vivo studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.