Learn More
Medchemexpress LLC Arecaidine hydrochloride | 6018-28-6 | 1 ML
SDP
Catalog No. 5000420273
Encompass_Preferred
Arecaidine hydrochloride is a pyridine alkaloid. It functions as a potent GABA uptake inhibitor and acts as a substrate of H+-coupled amino acid transporter 1 (PAT1, SLC36A1), competitively inhibiting L-proline uptake.
  • Pyridine alkaloid
  • Potent GABA uptake inhibitor
  • Functions as a substrate of H+-coupled amino acid transporter 1 (PAT1, SLC36A1)
  • Competitively inhibits L-proline uptake
  • Molecular formula: C7H12ClNO2
  • Derived from plants, specifically Palmae Areca catechu L.
Catalog No. 50-004-20273 Supplier Medchemexpress LLC Supplier No. HYN2368A10MM/1ML
Arecaidine hydrochloride is a pyridine alkaloid. It functions as a potent GABA uptake inhibitor and acts as a substrate of H+-coupled amino acid transporter 1 (PAT1, SLC36A1), competitively inhibiting L-proline uptake.
  • Pyridine alkaloid
  • Potent GABA uptake inhibitor
  • Functions as a substrate of H+-coupled amino acid transporter 1 (PAT1, SLC36A1)
  • Competitively inhibits L-proline uptake
  • Molecular formula: C7H12ClNO2
  • Derived from plants, specifically Palmae Areca catechu L.