Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arecaidine hydrochloride | 6018-28-6 | MFCD00051993 | 98.0% | 177.63 g·mol⁻1 | C7H12ClNO2 | 10 MG
SDP

Catalog No. 5000289535
Encompass_Preferred

Arecaidine hydrochloride is the hydrochloride salt of arecaidine, a pyridine alkaloid that acts as a GABA uptake inhibitor and a substrate of H+-coupled amino acid transporters. Supplied as a solid for research use, it is listed with high purity and recommended storage conditions.

  • Acts as a GABA uptake inhibitor and transporter substrate.
  • Molecular formula: C7H12ClNO2.
  • Molecular weight: 177.63 g·mol⁻1.
  • CAS number: 6018-28-6.
  • Purity: 98.0%.
  • Appearance: white to light yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • SMILES: O=C(C1=CCCN(C)C1)O.[H]Cl.
  • Available in 10 mg pack sizes for research use.

Catalog No. 50-002-89535 Supplier Medchemexpress LLC Supplier No. HYN2368A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arecaidine hydrochloride is the hydrochloride salt of arecaidine, a pyridine alkaloid that acts as a GABA uptake inhibitor and a substrate of H+-coupled amino acid transporters. Supplied as a solid for research use, it is listed with high purity and recommended storage conditions.

  • Acts as a GABA uptake inhibitor and transporter substrate.
  • Molecular formula: C7H12ClNO2.
  • Molecular weight: 177.63 g·mol⁻1.
  • CAS number: 6018-28-6.
  • Purity: 98.0%.
  • Appearance: white to light yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • SMILES: O=C(C1=CCCN(C)C1)O.[H]Cl.
  • Available in 10 mg pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.