Learn More
Selleck Chemical LLC Arecaidine hydrochloride
SDP
Catalog No. 503137033
Encompass_Preferred
Arecaidine hydrochloride a constituent of the nut of Areca catechu inhibits the uptake of GABA and beta-alanine but not that of glycine
Catalog No. 50-313-7033 Supplier Selleck Chemical LLC Supplier No. E05995MG
Arecaidine hydrochloride a constituent of the nut of Areca catechu inhibits the uptake of GABA and beta-alanine but not that of glycine