Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arecaidine propargyl ester hydrobromide | 116511-28-5 | MFCD00078566 | 99.5% | 260.13 g/mol | C10H14BrNO2 | 5 MG
SDP

Catalog No. 5000262709
Encompass_Preferred

Arecaidine propargyl ester hydrobromide is a research-grade small molecule that acts as an agonist of M2 muscarinic acetylcholine receptors and has been reported to induce apoptosis, increase reactive oxygen species, and cause G2/M cell-cycle arrest in certain tumor cell models.

  • Purity: 99.5%.
  • Appearance: white to off-white solid.
  • Molecular weight: 260.13 g/mol.
  • Chemical formula: C10H14BrNO2.
  • Biological activity: M2 muscarinic receptor agonist; reported inhibition of tumor cell proliferation.
  • Storage: store sealed at 4°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Pack sizes: available in 1 mg, 5 mg, and 10 mg.

Catalog No. 50-002-62709 Supplier Medchemexpress LLC Supplier No. HY1340615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arecaidine propargyl ester hydrobromide is a research-grade small molecule that acts as an agonist of M2 muscarinic acetylcholine receptors and has been reported to induce apoptosis, increase reactive oxygen species, and cause G2/M cell-cycle arrest in certain tumor cell models.

  • Purity: 99.5%.
  • Appearance: white to off-white solid.
  • Molecular weight: 260.13 g/mol.
  • Chemical formula: C10H14BrNO2.
  • Biological activity: M2 muscarinic receptor agonist; reported inhibition of tumor cell proliferation.
  • Storage: store sealed at 4°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Pack sizes: available in 1 mg, 5 mg, and 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.