Learn More
Medchemexpress LLC Arecaidine propargyl ester hydrobromide | 116511-28-5 | MFCD00078566 | 99.5% | C10H14BrNO2 | 10MG
SDP
Catalog No. 5000260422
Encompass_Preferred
Arecaidine propargyl ester hydrobromide is a small-molecule M2 muscarinic acetylcholine receptor agonist supplied as the hydrobromide salt. It has been reported to modulate M2 receptor activity and to inhibit tumor cell proliferation in vitro. The compound is provided as a white to off-white solid for laboratory research applications and is characterized by a high-purity specification.
Catalog No. 50-002-60422 Supplier Medchemexpress LLC Supplier No. HY13406110MG
Arecaidine propargyl ester hydrobromide is a small-molecule M2 muscarinic acetylcholine receptor agonist supplied as the hydrobromide salt. It has been reported to modulate M2 receptor activity and to inhibit tumor cell proliferation in vitro. The compound is provided as a white to off-white solid for laboratory research applications and is characterized by a high-purity specification.