Learn More
TARGETMOL CHEMICALS INC Arecoline 500MG
SDP
Catalog No. 502880131
Encompass_Preferred
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Arecoline is a natural alkaloid derived from areca nut in the palm family and is a potent nicotine and partial agonist of muscarinic acetylcholine receptors. Arecoline has anti-anxiety and anti-parasitic activity, can induce oxidative stress and can be used to study Alzheimer's disease and Alzheimer's disease. Purity 99.28%
Catalog No. 50-288-0131 Supplier TARGETMOL CHEMICALS INC Supplier No. T68953500MG
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Arecoline is a natural alkaloid derived from areca nut in the palm family and is a potent nicotine and partial agonist of muscarinic acetylcholine receptors. Arecoline has anti-anxiety and anti-parasitic activity, can induce oxidative stress and can be used to study Alzheimer's disease and Alzheimer's disease. Purity 99.28%