Learn More
Selleck Chemical LLC Arecoline HBr S2614-50mg
SDP
Catalog No. 503112522
Encompass_Preferred
Arecoline(NSC-31750) is a muscarinic acetylcholine receptor agonist
Catalog No. 50-311-2522 Supplier Selleck Chemical LLC Supplier No. S261450MG
Arecoline(NSC-31750) is a muscarinic acetylcholine receptor agonist