Learn More
Selleck Chemical LLC Arecoline HBr S2614-1g
SDP
Catalog No. 503112876
Encompass_Preferred
Arecoline(NSC-31750) is a muscarinic acetylcholine receptor agonist
Catalog No. 50-311-2876 Supplier Selleck Chemical LLC Supplier No. S26141G
Arecoline(NSC-31750) is a muscarinic acetylcholine receptor agonist