Learn More
Apexbio Technology LLC Arecoline hydrobromide 300-08-3 1g
SDP
Catalog No. 5000094498
Encompass_Preferred
Arecoline hydrobromide (CAS 300-08-3) is a nicotinic alkaloid that acts as a partial agonist at muscarinic acetylcholine receptors M1 M4 In electrophysiological studies it significantly inhibits the hERG potassium channel current with an IC50 of 9 55 mol/L The compound s inhibitory effect on IhERG displays concentration time and voltage dependency with blockade intensity influenced by stimulation frequency Subcutaneous administration in mice yields an LD50 of 100 mg/kg Arecoline hydrobromide is utilized in research on cardiac electrophysiology and muscarinic receptor pharmacology particularly for investigating frequency- and state-dependent modulation of cardiac ion channels
Catalog No. 50-000-94498 Supplier Apexbio Technology LLC Supplier No. B33701000
Arecoline hydrobromide (CAS 300-08-3) is a nicotinic alkaloid that acts as a partial agonist at muscarinic acetylcholine receptors M1 M4 In electrophysiological studies it significantly inhibits the hERG potassium channel current with an IC50 of 9 55 mol/L The compound s inhibitory effect on IhERG displays concentration time and voltage dependency with blockade intensity influenced by stimulation frequency Subcutaneous administration in mice yields an LD50 of 100 mg/kg Arecoline hydrobromide is utilized in research on cardiac electrophysiology and muscarinic receptor pharmacology particularly for investigating frequency- and state-dependent modulation of cardiac ion channels