Learn More
Medchemexpress LLC Arecoline hydrobromide | 300-08-3 | 99.9% | 100 MG
SDP
Catalog No. 5000407819
Encompass_Preferred
Arecoline hydrobromide is a naturally brain-penetrant and orally active psychoactive alkaloid. It functions as a partial agonist of nicotinic and muscarinic acetylcholine receptors, exhibiting stimulation, alertness, anxiolysis, and anti-parasitic effects. It can also induce oxidative stress.
  • Acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors
  • Exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects
  • Induces generation of reactive oxygen species and cell cycle arrest in HaCaT cells
  • Up-regulates stress-responsive genes
  • Alleviates anxiety and depression in CFA-induced mice
Catalog No. 50-004-07819 Supplier Medchemexpress LLC Supplier No. HYB0489100MG
Arecoline hydrobromide is a naturally brain-penetrant and orally active psychoactive alkaloid. It functions as a partial agonist of nicotinic and muscarinic acetylcholine receptors, exhibiting stimulation, alertness, anxiolysis, and anti-parasitic effects. It can also induce oxidative stress.
  • Acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors
  • Exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects
  • Induces generation of reactive oxygen species and cell cycle arrest in HaCaT cells
  • Up-regulates stress-responsive genes
  • Alleviates anxiety and depression in CFA-induced mice