Learn More
Medchemexpress LLC Arecoline hydrobromide | 300-08-3 | 99.95% | 1 ML
SDP
Catalog No. 5000407832
Encompass_Preferred
Arecoline hydrobromide is a naturally brain-penetrant and orally active psychoactive alkaloid. It acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors, exhibiting properties such as stimulation, alertness, anxiolysis, and anti-parasitic effects. It can also induce oxidative stress. This compound is for research use only.
  • Acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors
  • Exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects
  • Can induce oxidative stress
  • Brain-penetrant and orally active
  • Induces reactive oxygen species generation and G1/G0 phase cell cycle arrest in HaCaT cells
  • Upregulates stress-responsive genes
  • Alleviates anxiety and depression in CFA-induced mice
Catalog No. 50-004-07832 Supplier Medchemexpress LLC Supplier No. HYB048910MM/1ML
Arecoline hydrobromide is a naturally brain-penetrant and orally active psychoactive alkaloid. It acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors, exhibiting properties such as stimulation, alertness, anxiolysis, and anti-parasitic effects. It can also induce oxidative stress. This compound is for research use only.
  • Acts as a partial agonist of nicotinic and muscarinic acetylcholine receptors
  • Exhibits stimulation, alertness, anxiolysis, and anti-parasitic effects
  • Can induce oxidative stress
  • Brain-penetrant and orally active
  • Induces reactive oxygen species generation and G1/G0 phase cell cycle arrest in HaCaT cells
  • Upregulates stress-responsive genes
  • Alleviates anxiety and depression in CFA-induced mice