Learn More
Abcam Arecoline hydrobromide, Muscarinic receptor agonist, 250MG
Catalog No. 502706953
Encompass_Preferred
MW 236.11 Da, Purity >98%. Muscarinic receptor agonist (EC₅₀ values are 7 (M₁), 95 (M₂), 11 (M₃), 410 (M₄) and 69 nM (M₅)). Natural alkaloid. Induces decreased blood-brain barrier permeability.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6953 Supplier Abcam Supplier No. AB141044250MG
MW 236.11 Da, Purity >98%. Muscarinic receptor agonist (EC₅₀ values are 7 (M₁), 95 (M₂), 11 (M₃), 410 (M₄) and 69 nM (M₅)). Natural alkaloid. Induces decreased blood-brain barrier permeability.
The product is subject to the following: Abcam Restricted Use Statement