Learn More
Cayman Chemical Arecoline (hydrobromide), Purity: ≥95%, CAS: 300-08-3, 100 mg
SDP
Catalog No. 502955148
Encompass_Preferred
A natural alkaloid in the betel nut; an agonist of the muscarinic acetylcholine receptors M1, M2, M3, M4, and M5; causes smooth muscle contraction; has been shown to improve learning and memory and may prove to be useful in treating dementia
Catalog No. 50-295-5148 Supplier Cayman Chemical Supplier No. 13662100 MG
A natural alkaloid in the betel nut; an agonist of the muscarinic acetylcholine receptors M1, M2, M3, M4, and M5; causes smooth muscle contraction; has been shown to improve learning and memory and may prove to be useful in treating dementia