Learn More
Cayman Chemical Arenobufagin, Purity: ≥95%, CAS: 464-74-4, 10 mg
SDP
Catalog No. 502990363
Encompass_Preferred
A cardiotonic steroid with anticancer activity; inhibits Na+/K+-ATPase activity in isolated guinea pig ventricular myocytes (IC50 = 290 nM); increases cleavage of PARP, caspase-3, and caspase-9, as well as induces apoptosis in HepG2 hepatocellular carcinoma cells; It inhibits proliferation of HepG2, Hep3B, BEL-7402, and MCF-7 cancer cells (IC50s = 101.9, 39.8, 416.3, and 143.7 nM, respectively), as well as multidrug-resistant HepG2/adm and MCF-7/adr cancer cells (IC50s = 730 and 67.5 nM, respectively); reduces tumor growth, the number and size of lung metastases, and tumor levels of the EMT markers vimentin and β-catenin in a PC3 mouse xenograft model at 1 mg/kg; inhibits VEGF-induced angiogenesis in a Matrigel™ plug assay in mice at 5 and 10 µM/plug
Catalog No. 50-299-0363 Supplier Cayman Chemical Supplier No. 3069510 MG
A cardiotonic steroid with anticancer activity; inhibits Na+/K+-ATPase activity in isolated guinea pig ventricular myocytes (IC50 = 290 nM); increases cleavage of PARP, caspase-3, and caspase-9, as well as induces apoptosis in HepG2 hepatocellular carcinoma cells; It inhibits proliferation of HepG2, Hep3B, BEL-7402, and MCF-7 cancer cells (IC50s = 101.9, 39.8, 416.3, and 143.7 nM, respectively), as well as multidrug-resistant HepG2/adm and MCF-7/adr cancer cells (IC50s = 730 and 67.5 nM, respectively); reduces tumor growth, the number and size of lung metastases, and tumor levels of the EMT markers vimentin and β-catenin in a PC3 mouse xenograft model at 1 mg/kg; inhibits VEGF-induced angiogenesis in a Matrigel™ plug assay in mice at 5 and 10 µM/plug