Learn More
Selleck Chemical LLC Arenobufagin S0943-1MG
SDP
Catalog No. 5000041831
Encompass_Preferred
Arenobufagin is a natural bufadienolide from toad venom and induces apoptosis and autophagy in human hepatocellular carcinoma cells through inhibition of PI3K/Akt/mTOR pathway
Catalog No. 50-000-41831 Supplier Selleck Chemical LLC Supplier No. S09431MG
Arenobufagin is a natural bufadienolide from toad venom and induces apoptosis and autophagy in human hepatocellular carcinoma cells through inhibition of PI3K/Akt/mTOR pathway