Learn More
Chemglass Life Sciences AREX 5 Advance Magnetic Hot Plate Stirrer KIT with Probe, Vertical Support Rod, Boss Head, and Support/Holding Rod
SDP
Catalog No. 5000097375
Encompass_Preferred
State-of-the-art heating magnetic stirrer combining powerful performance advanced safety features several connectivity options and enhanced user experience Designed to meet the highest demands it ensures precision efficiency and uncompromising safety in every experiment Stirring Capacity Up to 20L (H2O)Speed Range 30 1 700RPMElevating Laboratory StandardsSet temperature ramps monitor viscosity switch stirring modes and streamline workflows with advanced timing and connectivity Remote control and monitoring via VELP Ermes Cloud Platform Save and transfer data via USB Precise ThermoregulationMaximum control for temperature-sensitive tasks with PT100 PT1000 probe or VTF Digital Thermoregulator When connected to probes two modes available Fast (rapid heating) and Fine (minimized overshoot) Thermoregulation range ambient to 310 C Advanced StirringDurable brushless motor with strong magnetic coupling Stirs up to 20 L from 30 to 1700 rpm
Catalog No. 50-000-97375 Supplier Chemglass Life Sciences Supplier No. CG1997V650
State-of-the-art heating magnetic stirrer combining powerful performance advanced safety features several connectivity options and enhanced user experience Designed to meet the highest demands it ensures precision efficiency and uncompromising safety in every experiment Stirring Capacity Up to 20L (H2O)Speed Range 30 1 700RPMElevating Laboratory StandardsSet temperature ramps monitor viscosity switch stirring modes and streamline workflows with advanced timing and connectivity Remote control and monitoring via VELP Ermes Cloud Platform Save and transfer data via USB Precise ThermoregulationMaximum control for temperature-sensitive tasks with PT100 PT1000 probe or VTF Digital Thermoregulator When connected to probes two modes available Fast (rapid heating) and Fine (minimized overshoot) Thermoregulation range ambient to 310 C Advanced StirringDurable brushless motor with strong magnetic coupling Stirs up to 20 L from 30 to 1700 rpm