Learn More
Enzo Life Sciences ARF inhibitory peptide (P-13) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLG4450001
Encompass_Preferred
Sequence is ADP-ribosylation factor (ARF) mARF1p (2-17) N-terminal peptide. Peptide inhibits ARF-dependent ADP-ribosylation by cholera toxin, endoplasmic reticulum to Golgi transport, intra-Golgi transport and endosome-endosome fusion in vitro at IC50=10-40 µM. Purity: ≥97%. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0885