Learn More
Aviva Systems Biology Corp ARF1 Antibody - middle region (ARP48184_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48184_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARF1. Alternate names: -ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish, Yeast
Catalog No. 50-014-59746