Learn More
Medchemexpress LLC ARF1 Human His-GST 20ug
SDP
Catalog No. 5000195758
Encompass_Preferred
Recombinant human ADP-ribosylation factor 1 (ARF1) supplied as a lyophilized, tagged protein for biochemical and in vitro assays. The product contains residues G2-K181, is expressed in E. coli, and is provided in a trehalose-stabilized formulation to support handling and storage.
Catalog No. 50-001-95758 Supplier Medchemexpress LLC Supplier No. HYP7208920UG
Recombinant human ADP-ribosylation factor 1 (ARF1) supplied as a lyophilized, tagged protein for biochemical and in vitro assays. The product contains residues G2-K181, is expressed in E. coli, and is provided in a trehalose-stabilized formulation to support handling and storage.