Learn More
Aviva Systems Biology Corp Arf4 Antibody - N-terminal region (ARP56032_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56032_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Arf4. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AA407803ApplicationWB. Species reactivity: Horse, Human, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-286-111