Learn More
Aviva Systems Biology Corp ARF6 Antibody - middle region (ARP54598_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54598_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARF6. Alternate names: DKFZp564M0264ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61253