Learn More
Aviva Systems Biology Corp ARFGAP1 Antibody - middle region (ARP57150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARFGAP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ARFGAP1. Alternate names: ARF1GAP, HRIHFB2281, MGC39924ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-286-268