Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arfolitixorin (calcium) | 1148151-21-6 | 95.9% | 495.50 g/mol | C20H21CaN7O6 | 5 MG
SDP

Catalog No. 5000263740
Encompass_Preferred

Arfolitixorin (calcium) is the calcium salt of arfolitixorin, an active folate derivative that functions as a 5-fluorouracil moderator and enzyme substrate. It is supplied as a solid reference standard for biochemical, pharmacological, and molecular biology studies requiring the active form of 5,10-methylenetetrahydrofolate.

  • Available in small vial pack sizes (1 mg, 5 mg, 10 mg).
  • Purity determined by HPLC: 95.9%.
  • Molecular formula C20H21CaN7O6 and molecular weight 495.50 g/mol.
  • Supplied as a research reagent for biochemical and pharmacological assays.
  • Used as a direct-acting folate to modulate 5-FU activity in research models.

Catalog No. 50-002-63740 Supplier Medchemexpress LLC Supplier No. HY109051A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Arfolitixorin (calcium) is the calcium salt of arfolitixorin, an active folate derivative that functions as a 5-fluorouracil moderator and enzyme substrate. It is supplied as a solid reference standard for biochemical, pharmacological, and molecular biology studies requiring the active form of 5,10-methylenetetrahydrofolate.

  • Available in small vial pack sizes (1 mg, 5 mg, 10 mg).
  • Purity determined by HPLC: 95.9%.
  • Molecular formula C20H21CaN7O6 and molecular weight 495.50 g/mol.
  • Supplied as a research reagent for biochemical and pharmacological assays.
  • Used as a direct-acting folate to modulate 5-FU activity in research models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.