Learn More
ABclonal Technology ARFRP1 / ARP Rabbit mAb
SDP
Catalog No. 5000008141
Encompass_Preferred
The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes Alternatively spliced transcript variants encoding different isoforms have been identified
Catalog No. 50-000-08141 Supplier ABclonal Technology Supplier No. A20884100UL
The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes Alternatively spliced transcript variants encoding different isoforms have been identified