Learn More
ABclonal Technology ARFRP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14311500UL
Encompass_Preferred
The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins. This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes. Alternatively spliced transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-09210