Learn More
Medchemexpress LLC (Arg)9, FAM-labeled | 1782.02 | 5 MG
SDP
Catalog No. 5000424486
Encompass_Preferred
(Arg)9, FAM-labeled is a cell-penetrating peptide (CPP), specifically a nona-arginine (ARG) with a FAM label. CPPs are used for delivering bioactive cargoes into the cytosol of intact cells.
  • Cell-penetrating peptide (CPP)
  • Nona-arginine (ARG) with FAM label
  • Delivers bioactive cargoes into the cytosol of intact cells
Catalog No. 50-004-24486 Supplier Medchemexpress LLC Supplier No. HYP25005MG
(Arg)9, FAM-labeled is a cell-penetrating peptide (CPP), specifically a nona-arginine (ARG) with a FAM label. CPPs are used for delivering bioactive cargoes into the cytosol of intact cells.
  • Cell-penetrating peptide (CPP)
  • Nona-arginine (ARG) with FAM label
  • Delivers bioactive cargoes into the cytosol of intact cells