Learn More
Medchemexpress LLC Arg-Arg-AMC acetate | 487.56 | 1 MG
SDP
Catalog No. 5000396648
Encompass_Preferred
Arg-Arg-AMC acetate is the acetate salt form of Arg-Arg-AMC. It functions as a fluorescent substrate for cathepsin B and is utilized in cathepsin B activity assays.
  • Fluorescent substrate for cathepsin B
  • Used in cathepsin B activity assays
  • Acetate salt form
Catalog No. 50-003-96648 Supplier Medchemexpress LLC Supplier No. HYP4340A1MG
Arg-Arg-AMC acetate is the acetate salt form of Arg-Arg-AMC. It functions as a fluorescent substrate for cathepsin B and is utilized in cathepsin B activity assays.
  • Fluorescent substrate for cathepsin B
  • Used in cathepsin B activity assays
  • Acetate salt form