Learn More
Medchemexpress LLC Arg-Glu(edans)-Ile-His-Pro-Phe-His-Pro-Phe-His-Leu-Val-Ile-His-Thr-Lys(dabcyl)-Arg TFA | 98.8% | 2778.12 | 5 MG
SDP
Catalog No. 5000398249
Encompass_Preferred
Arg-Glu(edans)-Ile-His-Pro-Phe-His-Pro-Phe-His-Leu-Val-Ile-His-Thr-Lys(dabcyl)-Arg TFA is a fluorogenic renin substrate based on the N-terminal tetradecapeptide sequence of human angiotensinogen (hTDP).
  • Fluorogenic renin substrate
  • Based on N-terminal tetradecapeptide sequence
  • Derived from human angiotensinogen (hTDP)
Catalog No. 50-003-98249 Supplier Medchemexpress LLC Supplier No. HYP5123A5MG
Arg-Glu(edans)-Ile-His-Pro-Phe-His-Pro-Phe-His-Leu-Val-Ile-His-Thr-Lys(dabcyl)-Arg TFA is a fluorogenic renin substrate based on the N-terminal tetradecapeptide sequence of human angiotensinogen (hTDP).
  • Fluorogenic renin substrate
  • Based on N-terminal tetradecapeptide sequence
  • Derived from human angiotensinogen (hTDP)