Learn More
Medchemexpress LLC Arg-Gly-Glu-Ser TFA | 99.5% | 561.47 | 5 MG
SDP
Catalog No. 5000425057
Encompass_Preferred
Arg-Gly-Glu-Ser TFA is a polypeptide that is related to RGD and functions as a negative control for Arg-Gly-Asp-Ser. In vivo studies have shown that this peptide does not significantly influence neutrophil and macrophage cell numbers or protein accumulation in comparison to LPS- or saline-treated mice.
  • Polypeptide related to RGD
  • Functions as a negative control for Arg-Gly-Asp-Ser
  • Does not influence neutrophil and macrophage cell numbers
  • No impact on protein accumulation compared to LPS- or saline-treated mice
Catalog No. 50-004-25057 Supplier Medchemexpress LLC Supplier No. HYP0309A5MG
Arg-Gly-Glu-Ser TFA is a polypeptide that is related to RGD and functions as a negative control for Arg-Gly-Asp-Ser. In vivo studies have shown that this peptide does not significantly influence neutrophil and macrophage cell numbers or protein accumulation in comparison to LPS- or saline-treated mice.
  • Polypeptide related to RGD
  • Functions as a negative control for Arg-Gly-Asp-Ser
  • Does not influence neutrophil and macrophage cell numbers
  • No impact on protein accumulation compared to LPS- or saline-treated mice